3-Bromo-N-(3-chloropropyl)benzenesulfonamide 97%

Supplier: Ambeed
Danger

A138776-1G A138776-25G A138776-5G
77095-354EA 12.74 USD
77095-354 77095-356 77095-358
3-Bromo-N-(3-chloropropyl)benzenesulfonamide 97%
3-Bromo-N-(3-chloropropyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₉H₁₁BrClNO₂S
MW: 312.6111 g/mol
MDL Number: MFCD09258645
CAS Number: 929000-46-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR