1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A138464-1G A138464-250MG
77095-332EA 44.56 USD
77095-332 77095-334
1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid 97%
1-(4-Methoxybenzyl)-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
MW: 232.235 g/mol
MDL Number: MFCD08456928
CAS Number: 1105039-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR