4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Aminophenoxy)-6,7-dimethoxyquinoline, 4-((6,7-Dimethoxyquinolin-4-yl)oxy)aniline

A138452-10G A138452-1G A138452-250MG A138452-25G A138452-5G
77095-322EA 112.46 USD
77095-322 77095-324 77095-326 77095-328 77095-330
4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline 97%
4-[(6,7-Dimethoxyquinolin-4-yl)oxy]aniline

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₃
MW: 296.33 g/mol
MDL Number: MFCD19685633
CAS Number: 190728-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR