Ac-Tyr(Ac)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Acetamido-3-(4-acetoxyphenyl)propanoic acid, N-Ac-Tyr(Ac)-OH

A138427-1G A138427-5G
77095-320EA 19 USD
77095-320 77900-912
Ac-Tyr(Ac)-OH 95%
Ac-Tyr(Ac)-OH

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₅
MW: 265.3 g/mol
MDL Number: MFCD00038242
CAS Number: 17355-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR