N-Ethyl-4-methoxybenzylamine Hydrochloride 98%

Supplier: Ambeed
Warning

A138302-10G A138302-1G A138302-5G
77095-314EA 130.5 USD
77095-314 77095-316 77095-318
N-Ethyl-4-methoxybenzylamine Hydrochloride 98%
N-Ethyl-4-methoxybenzylamine Hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₆ClNO
MW: 201.69 g/mol
MDL Number: MFCD07108568
CAS Number: 90389-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR