3-Acetyl-4-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A138106-100MG A138106-10G A138106-1G A138106-250MG A138106-5G
77095-302EA 19.1 USD
77095-302 77095-304 77095-306 77095-308 77095-310
3-Acetyl-4-hydroxybenzoic acid 95%
3-Acetyl-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.15 g/mol
MDL Number: MFCD00532680
CAS Number: 16357-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR