2',3'-Difluoroacetophenone 95%

Supplier: Ambeed
Warning

A137821-1G A137821-25G
77095-290EA 6 USD
77095-290 77901-473
2',3'-Difluoroacetophenone 95%
2',3'-Difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MDL Number: MFCD00061274
CAS Number: 18355-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR