Biphenyl-3,3'-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A136778-100MG A136778-1G A136778-250MG A136778-5G A136778-25G
77095-252EA 11 USD
77095-252 77095-254 77095-256 77095-258 77900-579
Biphenyl-3,3'-dicarboxylic acid 98%
Biphenyl-3,3'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀O₄
MDL Number: MFCD00845987
CAS Number: 612-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR