3-Methyl-4-nitropyridine 98%

Supplier: Ambeed
Warning

A136305-250MG A136305-25G A136305-5G
77095-220EA 10 USD
77095-220 77095-222 77095-224
3-Methyl-4-nitropyridine 98%
3-Methyl-4-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD03095084
CAS Number: 1678-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR