2-Chloro-4,6-difluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A136246-100MG A136246-1G A136246-250MG
77095-214EA 19 USD
77095-214 77095-216 77095-218
2-Chloro-4,6-difluorobenzaldehyde 95%
2-Chloro-4,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
MDL Number: MFCD18415553
CAS Number: 1261493-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR