4-Chloro-8-nitroquinoline 97%

Supplier: Ambeed
Warning

A136125-1G A136125-250MG A136125-25G
77095-208EA 46.68 USD
77095-208 77095-210 77095-212
4-Chloro-8-nitroquinoline 97%
4-Chloro-8-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.6 g/mol
MDL Number: MFCD00457008
CAS Number: 23833-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR