WH-4-023 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dimethylphenyl (2,4-dimethoxyphenyl)(2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)carbamate

A135851-10MG A135851-1MG A135851-5MG
77095-198EA 102.91 USD
77095-198 77095-200 77095-202
WH-4-023 98%
WH-4-023

Precaución: for Research Use Only

Formula: C₃₂H₃₆N₆O₄
MW: 568.7 g/mol
MDL Number: MFCD28138072
CAS Number: 837422-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR