1-Acetyl-3-bromoindole 97%

Supplier: Ambeed
Warning

A135463-1G A135463-5G
77095-194EA 20.16 USD
77095-194 77095-196
1-Acetyl-3-bromoindole 97%
1-Acetyl-3-bromoindole

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO
MW: 238.08 g/mol
MDL Number: MFCD00159880
CAS Number: 66417-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR