1-Fluoro-3-iodobenzene 98% stabilized

Supplier: Ambeed
Warning

Synonyms: 3-Fluoroiodobenzene

Stabilization: Stabilized with Copper chip

A135177-100G A135177-10G A135177-25G A135177-500G A135177-5G
77095-160EA 97.61 USD
77095-160 77095-162 77095-164 77095-166 77095-168
1-Fluoro-3-iodobenzene 98% stabilized
1-Fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₄FI
MW: 222 g/mol
MDL Number: MFCD00001044
CAS Number: 1121-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR