3-Nitropyridine-2,4-diamine 95%

Supplier: Ambeed
Warning

A134568-100MG A134568-250MG
77095-146EA 21.22 USD
77095-146 77095-148
3-Nitropyridine-2,4-diamine 95%
3-Nitropyridine-2,4-diamine

Precaución: for Research Use Only

Formula: C₅H₆N₄O₂
MW: 154.13 g/mol
MDL Number: MFCD01646408
CAS Number: 24501-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR