2-(3,5-Dimethoxyphenyl)propan-2-ol 97%

Supplier: Ambeed
Warning

A134453-1G A134453-5G
77095-142EA 91.24 USD
77095-142 77095-144
2-(3,5-Dimethoxyphenyl)propan-2-ol 97%
2-(3,5-Dimethoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
MDL Number: MFCD02684175
CAS Number: 39507-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR