2-(Trifluoromethyl)-4H-1-benzopyran-4-one 95%

Supplier: Ambeed
Warning

A134360-250MG A134360-1G
77095-140EA 22 USD
77095-140 77901-281
2-(Trifluoromethyl)-4H-1-benzopyran-4-one 95%
2-(Trifluoromethyl)-4H-1-benzopyran-4-one

Precaución: for Research Use Only

Formula: C₁₀H₅F₃O₂
MDL Number: MFCD00761957
CAS Number: 151668-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR