Bzl-Pro-OH 98%

Supplier: Ambeed
Warning

Synonyms: (2S)-1-Benzylpyrrolidine-2-carboxylic acid, (S)-1-Benzylpyrrolidine-2-carboxylic acid

A134192-100G A134192-10G A134192-1G A134192-250MG A134192-25G A134192-5G
77095-126EA 819.02 USD
77095-126 77095-128 77095-130 77095-132 77095-134 77095-136
Bzl-Pro-OH 98%
Bzl-Pro-OH

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₂
MW: 205.3 g/mol
MDL Number: MFCD00235894
CAS Number: 31795-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR