WHI-P154 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-[(6,7-dimethoxy-4-quinazolinyl)amino]phenol, 2-Bromo-4-((6,7-dimethoxyquinazolin-4-yl)amino)phenol

A134082-10MG A134082-1MG A134082-50MG A134082-5MG A134082-100MG A134082-250MG
77095-118EA 39 USD
77095-118 77095-120 77095-122 77095-124 77685-160 77685-161
WHI-P154 98%
WHI-P154

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrN₃O₃
MW: 376.2 g/mol
MDL Number: MFCD02179366
CAS Number: 211555-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR