1-Boc-3-isopropylpiperazine 95%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-3-Isopropylpiperazine

A134026-1G A134026-250MG
77095-114EA 106.09 USD
77095-114 77095-116
1-Boc-3-isopropylpiperazine 95%
1-Boc-3-isopropylpiperazine

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MW: 228.3312 g/mol
MDL Number: MFCD08273958
CAS Number: 502649-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR