5-Methyl-2-nitro-6,7-dihydropyrazolo[1,5-a]pyrazin-4(5H)-one 95%

Supplier: Ambeed
Warning

A133980-100MG A133980-250MG
77095-102EA 55.17 USD
77095-102 77095-104
5-Methyl-2-nitro-6,7-dihydropyrazolo[1,5-a]pyrazin-4(5H)-one 95%
5-Methyl-2-nitro-6,7-dihydropyrazolo[1,5-a]pyrazin-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₈N₄O₃
MW: 196.16 g/mol
MDL Number: MFCD22690143
CAS Number: 1408327-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR