3-Chloro-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-fluorobenzeneboronic acid, (3-Chloro-4-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-4-fluorophenyl)borane

A133762-100G A133762-1G A133762-5G A133762-10G A133762-25G A133762-500G
77095-096EA 127 USD
77095-096 77095-098 77095-100 77566-606 77685-157 77685-158
3-Chloro-4-fluorophenylboronic acid 98%
3-Chloro-4-fluorophenylboronic acid

Precaución: For research use only.

Formula: C₆H₅BClFO₂
MDL Number: MFCD00051800
CAS Number: 144432-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR