Methyl 3-acetyl-4-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A133709-100MG A133709-10G A133709-1G A133709-250MG A133709-5G
77095-086EA 33.95 USD
77095-086 77095-088 77095-090 77095-092 77095-094
Methyl 3-acetyl-4-hydroxybenzoate 95%
Methyl 3-acetyl-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD06797942
CAS Number: 57009-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR