2-Acetyl-2-thiazoline 98%

Supplier: Ambeed
Danger

Synonyms: 1-(4,5-Dihydrothiazol-2-yl)ethanone

A133353-1G A133353-25G A133353-5G
77095-066EA 15.92 USD
77095-066 77095-068 77095-070
2-Acetyl-2-thiazoline 98%
2-Acetyl-2-thiazoline

Precaución: for Research Use Only

Formula: C₅H₇NOS
MW: 129.18 g/mol
MDL Number: MFCD00209496
CAS Number: 29926-41-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR