4-Methyloxazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A133348-100MG A133348-25G A133348-5G A133348-10G
77095-060EA 5.31 USD
77095-060 77095-062 77095-064 77685-153
4-Methyloxazole-5-carboxylic acid 97%
4-Methyloxazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.1000 g/mol
MDL Number: MFCD01571070
CAS Number: 2510-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR