5H-Benzo[4,5]thieno[3,2-c]carbazole 99%

Supplier: Ambeed
Warning

A133304-1G A133304-250MG A133304-5G
77095-046EA 25 USD
77095-046 77095-048 77095-050
5H-Benzo[4,5]thieno[3,2-c]carbazole 99%
5H-Benzo[4,5]thieno[3,2-c]carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁NS
MDL Number: MFCD22571687
CAS Number: 1255308-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR