T-5224 98%

Supplier: Ambeed
Warning

Synonyms: 3-(5-(4-(Cyclopentyloxy)-2-hydroxybenzoyl)-2-((3-oxo-2,3-dihydrobenzo[d]isoxazol-6-yl)methoxy)phenyl)propanoic acid

A132992-10MG A132992-1MG A132992-5MG A132992-50MG
77095-034EA 63.66 USD
77095-034 77095-036 77095-038 77685-152
T-5224 98%
T-5224

Precaución: for Research Use Only

Formula: C₂₉H₂₇NO₈
MW: 517.54 g/mol
MDL Number: MFCD28053483
CAS Number: 530141-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR