2-Bromopyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A132402-5G A132402-250MG
77095-022EA 5711.89 USD
77095-022 77685-148
2-Bromopyrimidine-5-carbonitrile 95%
2-Bromopyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 183.99 g/mol
MDL Number: MFCD14702744
CAS Number: 1209458-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR