tert-Butyl methyl(piperidin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A130568-250MG A130568-1G A130568-5G
77094-948EA 29 USD
77094-948 77685-136 77900-493
tert-Butyl methyl(piperidin-3-yl)carbamate 97%
tert-Butyl methyl(piperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD06656642
CAS Number: 172478-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR