8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 97%

Supplier: Ambeed
Warning

A130188-100MG A130188-1G A130188-250MG
77094-930EA 19.1 USD
77094-930 77094-932 77094-934
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 97%
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one

Precaución: for Research Use Only

Formula: C₁₁H₉FN₂O
MW: 204.2 g/mol
MDL Number: MFCD23381312
CAS Number: 1408282-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR