Diethyl [(4-methylbenzenesulfonyl)methyl]-phosphonate 95%

Supplier: Ambeed
Warning

A129334-100MG A129334-1G A129334-250MG A129334-25G A129334-5G
77094-916EA 5 USD
77094-916 77094-918 77094-920 77094-922 77094-924
Diethyl [(4-methylbenzenesulfonyl)methyl]-phosphonate 95%

Precaución: for Research Use Only

Formula: C₁₂H₁₉O₅PS
MDL Number: MFCD28369627
CAS Number: 60682-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR