2-Chloro-4-fluorophenylhydrazine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-Chloro-4-fluorophenyl)hydrazine hydrochloride
A128943-100G
A128943-10G
A128943-1G
A128943-25G
A128943-5G
77094-890EA
151
USD
77094-890
77094-892
77094-894
77094-896
77094-898
2-Chloro-4-fluorophenylhydrazine hydrochloride 98%
2-Chloro-4-fluorophenylhydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₇Cl₂FN₂ |
MDL Number:
MFCD03788524 CAS Number: 497959-29-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...