1-Methyl-2-piperidinemethanol 98%

Supplier: Ambeed
Warning

A128852-100G A128852-5G
77094-884EA 71.09 USD
77094-884 77094-888
1-Methyl-2-piperidinemethanol 98%
1-Methyl-2-piperidinemethanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD00006494
CAS Number: 20845-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR