9-(4-Biphenylyl)carbazole 97%

Supplier: Ambeed
Warning

A128729-1G A128729-5G
77094-874EA 8.49 USD
77094-874 77094-876
9-(4-Biphenylyl)carbazole 97%
9-(4-Biphenylyl)carbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₇N
MW: 319.41 g/mol
MDL Number: MFCD28147704
CAS Number: 6299-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR