3-Fluoro-4-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Fluoropyridine-4-boronic acid pinacol ester

A127718-10G A127718-1G A127718-250MG A127718-5G
77094-838EA 82 USD
77094-838 77094-840 77094-842 77094-844
3-Fluoro-4-pyridineboronic acid pinacol ester 98%
3-Fluoro-4-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₂
MDL Number: MFCD06798251
CAS Number: 458532-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR