4-nitrophenyl 2-(pyridin-2-yldisulfanyl)ethyl carbonate 95%

Supplier: Ambeed
Warning

A127653-100MG A127653-1G
77094-836EA 50 USD
77094-836 77313-248
4-nitrophenyl 2-(pyridin-2-yldisulfanyl)ethyl carbonate 95%
4-nitrophenyl 2-(pyridin-2-yldisulfanyl)ethyl carbonate

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₅S₂
MDL Number: MFCD28556879
CAS Number: 874302-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR