DL-Asparagine monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: DL-Aspartic acid 4-amide monohydrate, H-DL-Asn-OH monohydrate, (±)-2-Aminosuccinic acid 4-amide monohydrate, DL-Asparagine hydrate, H-DL-Asn-OH·H₂O, DL-2-Aminosuccinamic acid monohydrate, 2,4-Diamino-4-oxobutanoic acid monohydrate

A127546-100G A127546-10G A127546-25G
77094-832EA 29.71 USD
77094-832 77094-834 77313-218
DL-Asparagine monohydrate 98%
DL-Asparagine monohydrate

Precaución: for Research Use Only

Formula: C₄H₈N₂O₃·H₂O
MW: 150.13 g/mol
MDL Number: MFCD00151039
CAS Number: 69833-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR