4'-Ethylbiphenyl-4-ylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4'-Ethyl-[1,1'-biphenyl]-4-yl)boronic acid

A127185-1G A127185-250MG A127185-5G
77094-818EA 26.53 USD
77094-818 77094-820 77685-105
4'-Ethylbiphenyl-4-ylboronic acid 97%
4'-Ethylbiphenyl-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BO₂
MW: 226.0800 g/mol
MDL Number: MFCD04038068
CAS Number: 153035-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR