3-Methylpiperidin-4-one hydrochloride 97%

Supplier: Ambeed
Warning

A126911-10G A126911-1G A126911-250MG A126911-25G A126911-5G
77094-802EA 163.38 USD
77094-802 77094-804 77094-806 77094-808 77094-810
3-Methylpiperidin-4-one hydrochloride 97%
3-Methylpiperidin-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MW: 149.62 g/mol
MDL Number: MFCD07369967
CAS Number: 4629-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR