Sodium 5-(2-carbamoylhydrazono)-1-methyl-6-oxo-2,3,5,6-tetrahydro-1H-indole-2-sulfonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Carbazochrome sodium sulphonate
A126021-100G
A126021-1G
A126021-250MG
A126021-25G
A126021-5G
77094-776EA
325.7
USD
77094-776
77094-778
77094-780
77094-782
77094-784
Sodium 5-(2-carbamoylhydrazono)-1-methyl-6-oxo-2,3,5,6-tetrahydro-1H-indole-2-sulfonate 98%
Sodium 5-(2-carbamoylhydrazono)-1-methyl-6-oxo-2,3,5,6-tetrahydro-1H-indole-2-sulfonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁N₄NaO₅S MW: 322.27 g/mol |
MDL Number:
MFCD00867493 CAS Number: 51460-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...