(3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate 95%

Supplier: Ambeed
Warning

A125843-10G A125843-25G A125843-5G
77094-762EA 15 USD
77094-762 77094-764 77094-766 77094-768 77094-770
(3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate 95%
(3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₁₁
MDL Number: MFCD00080787
CAS Number: 83-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR