Prasugrel 98%

Supplier: Ambeed
Warning

Synonyms: 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl Acetate, 5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate

A125835-100MG A125835-1MG A125835-25MG A125835-1G A125835-250MG A125835-5G
77094-756EA 11.67 USD
77094-756 77094-758 77094-760 77592-442 77611-074 77685-098
Prasugrel 98%
Prasugrel

Precaución: for Research Use Only

Formula: C₂₀H₂₀FNO₃S
MW: 373.44 g/mol
MDL Number: MFCD09954140
CAS Number: 150322-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR