5-Chloropyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A125826-100G A125826-10G A125826-1G A125826-250MG A125826-25G A125826-5G
77094-744EA 407.39 USD
77094-744 77094-746 77094-748 77094-750 77094-752 77094-754
5-Chloropyrazolo[1,5-a]pyrimidine 97%
5-Chloropyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.569 g/mol
MDL Number: MFCD04035683
CAS Number: 29274-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR