1,10-Dibromodecane 98%
Supplier: Ambeed
|
Warning
|
A125763-100G
A125763-500G
A125763-25G
A125763-10G
A125763-1000G
77094-734EA
43
USD
77094-734
77094-736
77312-874
77611-072
77685-096
1,10-Dibromodecane 98%
1,10-Dibromodecane
Precaución: for Research Use Only
Formula:
BrH₂C(CH₂)₈CH₂Br |
MDL Number:
MFCD00000222 CAS Number: 4101-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...