DL-Serine 98+%

Supplier: Ambeed
Warning

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

A125328-1000G A125328-100G A125328-25G A125328-500G
77094-722EA 85 USD
77094-722 77094-724 77094-726 77094-728
DL-Serine 98+%
DL-Serine

Precaución: for Research Use Only

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 395
MDL Number: MFCD00064223
CAS Number: 302-84-1
Merck Index: 13,08534

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR