Rutin 96%

Supplier: Ambeed
Warning

Synonyms: Ilixanthi, Myrticalorin, Globulariacitrin, Osyritrin, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4-oxo-4H-chromen-3-yl, Quercetin 3-rutinoside, Rutinoside, 3-Rutinosylquercetin, Quercetin 3-rhamnoglucoside, Paliuroside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, 3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside, Tanrutin, Phytomelin, Violaquercitrin, Sophorin, Rutoside

A125100-1000G A125100-100G A125100-10G A125100-25G A125100-500G
77094-710EA 173.99 USD
77094-710 77094-712 77094-714 77094-716 77094-718
Rutin 96%
Rutin

Precaución: for Research Use Only

Formula: C₂₇H₃₀O₁₆
MW: 610.521 g/mol
MDL Number: MFCD00006830
CAS Number: 153-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR