2-Fluoro-5-propoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-proproxyphenylboronic acid

A125014-1G A125014-250MG A125014-5G
77094-704EA 39.26 USD
77094-704 77094-706 77094-708
2-Fluoro-5-propoxyphenylboronic acid 98%
2-Fluoro-5-propoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₂BFO₃
MW: 197.99 g/mol
MDL Number: MFCD05664318
CAS Number: 863248-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR