6-Chloro-4-hydroxyquinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A124961-1G A124961-250MG
77094-702EA 190.97 USD
77094-702 77571-492
6-Chloro-4-hydroxyquinolin-2(1H)-one 95%
6-Chloro-4-hydroxyquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MW: 195.60 g/mol
MDL Number: MFCD04037761
CAS Number: 1677-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR