1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Allopurinol
A124580-100G
A124580-10G
A124580-1G
A124580-25G
A124580-5G
77094-688EA
29.71
USD
77094-688
77094-690
77094-692
77094-694
77094-696
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one 98%
1H-Pyrazolo[3,4-d]pyrimidin-4(5H)-one
Precaución: for Research Use Only
Formula:
C₅H₄N₄O MW: 136.11 g/mol |
MDL Number:
MFCD00599413 CAS Number: 315-30-0 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...