2-Methyl-4-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)butan-2-ol 95%

Supplier: Ambeed
Warning

A124553-100MG A124553-250MG
77094-684EA 31.83 USD
77094-684 77094-686
2-Methyl-4-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)butan-2-ol 95%
2-Methyl-4-(2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)butan-2-ol

Precaución: for Research Use Only

Formula: C₁₇H₂₇BO₃
MW: 290.20 g/mol
MDL Number: MFCD18734391
CAS Number: 1362243-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR